4-(4-Nitrophenyl)morpholine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
N1124-5G
N1124-25G
TCIAN1124-5GEA
16
EUR
TCIAN1124-5G
TCIAN1124-25G
4-(4-Nitrophenyl)morpholine ≥98.0% (by GC, titration analysis)
4-(4-Nitrophenyl)morpholine
Formula:
C₁₀H₁₂N₂O₃ |
MDL Number:
MFCD00023317 CAS Number: 10389-51-2 EINECS: 233-851-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 151.0 to 155.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...