Stampa…

exo-Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione, rel-(3aR,4S,7R,7aS)-Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione, exo-2,3-Norbornanedicarboximide

N1116-1G N1116-5G
TCIAN1116-1GEA 27 EUR
TCIAN1116-1G TCIAN1116-5G
exo-Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione ≥98.0% (by HPLC, titration analysis)
exo-Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione
Formula: C₉H₁₁NO₂
Peso molecolare: 165,1900 g/mol
MDL Number: MFCD12964181
CAS Number: 14805-29-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6f97737c8c710c8f232822d8c0a8b195427542a&vid=4cfdc01430d078f291ed69cfd8eb99fbe6bf1d50&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 151.0 to 155.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR