Stampa…

Neotame ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: (S)-3-((3,3-Dimethylbutyl)amino)-4-(((S)-1-methoxy-1-oxo-3-phenylpropan-2-yl)amino)-4-oxobutanoic acid, N-[N-(3,3-Dimethylbutyl)-L-alpha-aspartyl]-L-phenylalanine 1-Methyl ester, N-(N-(3,3-Dimethylbutyl)-L-alpha-aspartyl)-L-phenylalanine 1-methyl ester

N1112-1G N1112-25G N1112-5G
TCIAN1112-1GEA 16 EUR
TCIAN1112-1G TCIAN1112-25G TCIAN1112-5G
Neotame ≥98.0% (by HPLC, titration analysis)
Neotame
Formula: C₂₀H₃₀N₂O₅
Peso molecolare: 378,47 g/mol
MDL Number: MFCD09039056
CAS Number: 165450-17-9
Merck Index: 14,06465

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 84.0 to 88.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR