2-(Perfluorobutyl)ethyl acrylate ≥98.0% (by GC) stabilizzato
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3,3,4,4,5,5,6,6,6-Nonafluorohexyl Acrylate, Acrylic acid 3,3,4,4,5,5,6,6,6-Nonafluorohexyl ester, Acrylic acid 1H,1H,2H,2H-Nonafluorohexyl ester, 1H,1H,2H,2H-Nonafluorohexyl Acrylate
Stabilization:
stabilizzata con 4-ter-butil pirocatecolo
Formula:
C₉H₇F₉O₂ Peso molecolare: 318,139 g/mol |
MDL Number:
MFCD00236104 CAS Number: 52591-27-2 UN: 3272 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...