NVP-BHG712 ≥95.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
NVP-BHG 712, 4-Methyl-3-((1-methyl-6-(pyridin-3-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-yl)amino)-N-(3-(trifluoromethyl)phenyl)benzamide, 4-Methyl-3-[[1-methyl-6-(3-pyridinyl)-1H-pyrazolo[3,4-d]pyrimidin-4-yl]amino]-N-[3-(trifluoromethyl)phenyl]benzamide
Formula:
C₂₆H₂₀F₃N₇O Peso molecolare: 503,49 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD20272929 CAS Number: 940310-85-0 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Elemental analysis (Nitrogen) | 18.5 to 20.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...