Nonamethylene Glycol Dimethacrylate ≥98.0% (by GC) stabilizzato
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1,9-Nonanedioldimethacrylate, 1,9-Nonanediol Dimethacrylate
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
N1089-25G
N1089-500G
TCIAN1089-25GEA
23
EUR
TCIAN1089-25G
TCIAN1089-500G
Nonamethylene Glycol Dimethacrylate ≥98.0% (by GC) stabilizzato
Nonamethylene Glycol Dimethacrylate
Formula:
C₁₇H₂₈O₄ Peso molecolare: 296,41 g/mol |
MDL Number:
MFCD00080593 CAS Number: 65833-30-9 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...