Stampa…

Diamond nanoparticles 3% dispersion in ethylene glycol (Amine-modified) (<10 nm)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Nanodiamond

N1083-5G N1083-25G
TCIAN1083-5GEA 96 EUR
TCIAN1083-5G TCIAN1083-25G
Diamond nanoparticles 3% dispersion in ethylene glycol (Amine-modified) (<10 nm)
Diamond

Nanodiamonds (Nanocarbon Materials)

MDL Number: MFCD00211867
CAS Number: 7782-40-3

Visualizza i codici

Risultati del test di specificazione

Appearance Light gray to Brown to Black clear liquid to cloudy liquid
Concentration 2.5 to 3.4 %
Water max. 1.0 %
Mean particle diameter max. 10 nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR