Stampa…

4-Nitro-2-(trifluoromethyl)benzonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

N1068-5G N1068-25G
TCIAN1068-5GEA 57 EUR
TCIAN1068-5G TCIAN1068-25G
4-Nitro-2-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
4-Nitro-2-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₃N₂O₂
Peso molecolare: 216,12 g/mol
MDL Number: MFCD01075729
CAS Number: 320-47-8
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69cccc5fab66dbc71128f50f08135ac89ec2deaf&vid=69cccc5fab66dbc71128f50f08135ac89ec2deaf&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 49.0 to 53.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR