(S)-1-(Naphthalen-1-yl)ethanol ≥97.0% (by GC)
Fornitore: TCI
N1063-1G
N1063-5G
TCIAN1063-1GEA
38
EUR
TCIAN1063-1G
TCIAN1063-5G
(S)-1-(Naphthalen-1-yl)ethanol ≥97.0% (by GC)
(S)-1-(Naphthalen-1-yl)ethanol
Formula:
C₁₂H₁₂O Punto di ebollizione: 165°C |
MDL Number:
MFCD00077831 CAS Number: 15914-84-8 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 44.0 to 48.0 deg-C |
Specific rotation [a]20/D | -75.0 to -80.0 deg (C=1,MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...