Stampa…

nitro-Grela ≥97.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: [1,3-Bis(2,4,6-trimethylphenyl)imidazolidin-2-ylidene]-dichloro-[(5-nitro-2-propan-2-yloxyphenyl)methylidene]ruthenium, [1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro[(2-isopropoxy-5-nitrobenzylidene)]ruthenium(II)

N1060-100MG N1060-500MG
TCIAN1060-100MGEA 171 EUR
TCIAN1060-100MG TCIAN1060-500MG
nitro-Grela ≥97.0% (by HPLC)
nitro-Grela
Formula: C₃₁H₃₇Cl₂N₃O₃Ru
Peso molecolare: 671,63 g/mol
MDL Number: MFCD28411645
CAS Number: 502964-52-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2100ccccf774bcc9847d0c5c70c3998f2bb1010&vid=b2100ccccf774bcc9847d0c5c70c3998f2bb1010&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Green to Dark green powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Nitrogen) 5.90 to 6.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR