Stampa…

1-Naphthylmethylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Naphthalenemethylamine, Naphthalen-1-ylmethanamine, Naphthalene-1-methylamine, 1-(Aminomethyl)naphthalene

N1042-5G N1042-25G
TCIAN1042-5GEA 104 EUR
TCIAN1042-5G TCIAN1042-25G
1-Naphthylmethylamine ≥98.0% (by GC, titration analysis)
1-Naphthylmethylamine
Formula: C₁₁H₁₁N
Peso molecolare: 157,22 g/mol
Punto di ebollizione: 293°C
MDL Number: MFCD00004048
CAS Number: 118-31-0
EINECS: 204-244-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5eeb983afca47521bd9e9c0962297e98c6fa576b&vid=5eeb983afca47521bd9e9c0962297e98c6fa576b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR