4,4,5,5,6,6,7,7,7-Nonafluoro-1-heptanol ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
3-(Perfluorobutyl)-1-propanol, Daikin A-1430, 1H,1H,2H,2H,3H,3H-Nonafluoro-1-heptanol, 1H,1H,2H,2H,3H,3H-Perfluoro-1-heptanol, 4,4,5,5,6,6,7,7,7-Nonafluoroheptan-1-ol, 3-(Perfluorobutyl)propanol
N1040-5G
N1040-25G
N1040-100G
TCIAN1040-5GEA
65
EUR
TCIAN1040-5G
TCIAN1040-25G
TCIAN1040-100G
4,4,5,5,6,6,7,7,7-Nonafluoro-1-heptanol ≥98.0% (by GC)
4,4,5,5,6,6,7,7,7-Nonafluoro-1-heptanol
Formula:
C₇H₇F₉O Peso molecolare: 278,12 g/mol Punto di ebollizione: 64°C Flash Pt: 76°C |
MDL Number:
MFCD02183552 CAS Number: 83310-97-8 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...