Stampa…
Perfluoro(2-methyl-3-pentanone) ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms: 1,1,1,2,2,4,5,5,5-Nonafluoro-4-(trifluoromethyl)-3-pentanone, Perfluoroethyl Perfluoroisopropyl ketone
N1038-25G N1038-100G
TCIAN1038-25GEA 92 EUR
TCIAN1038-25G TCIAN1038-100G
Perfluoro(2-methyl-3-pentanone) ≥98.0% (by GC)
Perfluoro(2-methyl-3-pentanone)
Formula: C₆F₁₂O
Peso molecolare: 316,05 g/mol
MDL Number: MFCD04038343
CAS Number: 756-13-8
EINECS: 436-710-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eefc4ca97c1d259cee3a188b40b25504f8b01ba8&vid=eefc4ca97c1d259cee3a188b40b25504f8b01ba8&prid=web&clid=SAPDAM
Visualizza i codici
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %