Stampa…

4-Nitro-α,α,α-trifluoro-o-toluic acid ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Nitro-2-(trifluoromethyl)benzoic acid

N1037-1G N1037-5G
TCIAN1037-1GEA 27 EUR
TCIAN1037-1G TCIAN1037-5G
4-Nitro-α,α,α-trifluoro-o-toluic acid ≥97.0% (by GC, titration analysis)
4-Nitro-α,α,α-trifluoro-o-toluic acid
Formula: C₈H₄F₃NO₄
Peso molecolare: 235,12 g/mol
MDL Number: MFCD04039246
CAS Number: 320-37-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7be366b4d7ea302d9236e2b88709710ce83cbaf5&vid=7be366b4d7ea302d9236e2b88709710ce83cbaf5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 139.0 to 143.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR