Stampa…

Dimethylammonium dichlorotri(mu-chloro)bis{(R)-(+)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}diruthenate(II)

Fornitore: TCI EUROPE

Synonyms: [NH2Me2][{RuCl((R)-dm-segphos®)}2(mu-Cl)3]

N1023-200MG N1023-1G
TCIAN1023-200MGEA 38 EUR
TCIAN1023-200MG TCIAN1023-1G
Dimethylammonium dichlorotri(mu-chloro)bis{(R)-(+)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis{(R)-(+)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}diruthenate(II)
Formula: C₉₄H₉₆Cl₅NO₈P₄Ru₂
Peso molecolare: 1871,09 g/mol
MDL Number: MFCD09753038
CAS Number: 935449-46-0

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
IR to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR