Stampa…

Dimethylammonium dichlorotri(mu-chloro)bis{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}diruthenate(II)

Fornitore: TCI EUROPE

Synonyms: [NH2Me2][(RuCl((R)-xylbinap))2(mu-Cl)3], [NH2Me2][{RuCl((R)-xylbinap)}2(mu-Cl)3], Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl]diruthenate(II)

N1019-200MG N1019-1G
TCIAN1019-200MGEA 41 EUR
TCIAN1019-200MG TCIAN1019-1G
Dimethylammonium dichlorotri(mu-chloro)bis{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}diruthenate(II)
Formula: C₁₀₆H₁₀₄Cl₅NP₄Ru₂
Peso molecolare: 1895,29 g/mol
MDL Number: MFCD09753035
CAS Number: 944451-08-5

Visualizza i codici

Risultati del test di specificazione

Appearance Orange to Brown powder to crystal
IR to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR