Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II)
Fornitore: TCI
|
Attenzione
|
Formula:
C₉₀H₇₂Cl₅NP₄Ru₂ |
MDL Number:
MFCD09753033 CAS Number: 199684-47-4 |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
IR | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...