9-(1-Naphthyl)carbazole ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₂₂H₁₅N |
MDL Number:
MFCD26127424 CAS Number: 22034-43-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 126.0 to 131.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...