Stampa…

1-(4-Nitrobenzyl)-1H-1,2,4-triazole ≥98.0% (by GC)

Fornitore: TCI

Synonyms: 1-(4-Nitrobenzyl)-1,2,4-triazole

N1007-5G N1007-25G
TCIAN1007-5GEA 76 EUR
TCIAN1007-5G TCIAN1007-25G
1-(4-Nitrobenzyl)-1H-1,2,4-triazole ≥98.0% (by GC)
1-(4-Nitrobenzyl)-1H-1,2,4-triazole
Formula: C₉H₈N₄O₂
MDL Number: MFCD03840588
CAS Number: 119192-09-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=400cddb9c31e6e09d9170ead474159927cd99c55&vid=400cddb9c31e6e09d9170ead474159927cd99c55&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 98.0 %
Melting point 100.0 to 104.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR