Stampa…

3-Nitro-4-quinolinol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Hydroxy-3-nitroquinoline, 3-Nitroquinolin-4-ol

N0992-200MG N0992-1G
TCIAN0992-200MGEA 40 EUR
TCIAN0992-200MG TCIAN0992-1G
3-Nitro-4-quinolinol ≥98.0% (by GC, titration analysis)
3-Nitro-4-quinolinol
Formula: C₉H₆N₂O₃
Peso molecolare: 190,1600 g/mol
Punto di fusione: 361°C
MDL Number: MFCD00277722
CAS Number: 50332-66-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a490f5d98017ccc897e80ec51eb49d0917ec274&vid=2a490f5d98017ccc897e80ec51eb49d0917ec274&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR