Stampa…

N-(4-Nitrophenyl)-3-oxobutyramide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI

Synonyms: 4'-Nitroacetoacetanilide

N0990-200MG N0990-1G
TCIAN0990-200MGEA 79 EUR
TCIAN0990-200MG TCIAN0990-1G
N-(4-Nitrophenyl)-3-oxobutyramide ≥98.0% (by HPLC, titration analysis)
N-(4-Nitrophenyl)-3-oxobutyramide
Formula: C₁₀H₁₀N₂O₄
MDL Number: MFCD00452522
CAS Number: 4835-39-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a4c49461c1aa589ea285bca48016bff71ca2857&vid=6a4c49461c1aa589ea285bca48016bff71ca2857&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Amber powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 119.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR