1-Nitropyrazole ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
N0965-5G
N0965-25G
TCIAN0965-5GEA
55
EUR
TCIAN0965-5G
TCIAN0965-25G
1-Nitropyrazole ≥97.0% (by GC)
1-Nitropyrazole
Formula:
C₃H₃N₃O₂ Peso molecolare: 113,0748 g/mol |
MDL Number:
MFCD00015894 CAS Number: 7119-95-1 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 92.0 to 96.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...