Stampa…

3-(3-Nitrophenyl)propionic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Nitrohydrocinnamic acid

N0960-5G N0960-25G
TCIAN0960-5GEA 81 EUR
TCIAN0960-5G TCIAN0960-25G
3-(3-Nitrophenyl)propionic acid ≥98.0% (by GC, titration analysis)
3-(3-Nitrophenyl)propionic acid
Formula: C₉H₉NO₄
Peso molecolare: 195,1721 g/mol
MDL Number: MFCD01310835
CAS Number: 1664-57-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5c36c440073bf55ce51c8274598891f2f436119&vid=f5c36c440073bf55ce51c8274598891f2f436119&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 117.0 to 121.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR