Nafamostat mesylate ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
4-Guanidinobenzoic acid 6-Amidino-2-naphthyl Ester Bis(methanesulphonate), 6-Carbamimidoylnaphthalen-2-yl 4-guanidinobenzoate dimethanesulphonate, 6-Amidino-2-naphthyl 4-Guanidinobenzoate Bis(methanesulphonate)
N0959-20MG
N0959-100MG
TCIAN0959-20MGEA
35
EUR
TCIAN0959-20MG
TCIAN0959-100MG
Nafamostat mesylate ≥98.0% (by HPLC)
Nafamostat mesylate
Formula:
C₁₉H₁₇N₅O₂ Peso molecolare: 539,58 g/mol |
MDL Number:
MFCD00941430 CAS Number: 82956-11-4 Merck Index: 14,06350 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...