Nonanal diethyl acetal ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Nonyl Aldehyde Diethyl Acetal, 1,1-Diethoxynonane
N0956-5ML
N0956-25ML
TCIAN0956-5MLEA
29
EUR
TCIAN0956-5ML
TCIAN0956-25ML
Nonanal diethyl acetal ≥98.0% (by GC)
Nonanal diethyl acetal
Formula:
C₁₃H₂₈O₂ Peso molecolare: 216,37 g/mol Punto di ebollizione: 66°C |
MDL Number:
MFCD00672806 CAS Number: 54815-13-3 EINECS: 259-360-4 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...