Stampa…

3-(4-Nitrophenyl)propionic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Nitrohydrocinnamic Acid, 3-(4-Nitrophenyl)propanoic acid

N0944-1G N0944-5G
TCIAN0944-1GEA 30 EUR
TCIAN0944-1G TCIAN0944-5G
3-(4-Nitrophenyl)propionic acid ≥98.0% (by GC, titration analysis)
3-(4-Nitrophenyl)propionic acid
Formula: C₉H₉NO₄
Peso molecolare: 195,17 g/mol
MDL Number: MFCD00126834
CAS Number: 16642-79-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2736beee036ac4ffe395ac9dc2182d69e07aa30d&vid=2736beee036ac4ffe395ac9dc2182d69e07aa30d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 165.0 to 169.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR