Stampa…

5-Nitro-2-thiophenecarboxylic acid ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Nitrothiophene-2-carboxylic acid

N0940-5G N0940-25G
TCIAN0940-5GEA 48 EUR
TCIAN0940-5G TCIAN0940-25G
5-Nitro-2-thiophenecarboxylic acid ≥97.0% (by GC, titration analysis)
5-Nitro-2-thiophenecarboxylic acid
Formula: C₅H₃NO₄S
Peso molecolare: 173,14 g/mol
MDL Number: MFCD00159552
CAS Number: 6317-37-9
EINECS: 228-654-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a6eff30da500276942913503d144fe8b65f7fb0&vid=2a6eff30da500276942913503d144fe8b65f7fb0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 158.0 to 162.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR