Stampa…

1,4-Naphthalenebis(trifluoromethanesulphonate) ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1,4-Naphthaleneditriflate

N0934-1G N0934-5G
TCIAN0934-1GEA 94 EUR
TCIAN0934-1G TCIAN0934-5G
1,4-Naphthalenebis(trifluoromethanesulphonate) ≥98.0% (by GC)
1,4-Naphthalenebis(trifluoromethanesulphonate)
Formula: C₁₂H₆F₆O₆S₂
Peso molecolare: 424,28 g/mol
Temperatura di stoccaggio: Refrigerata
CAS Number: 152873-78-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR