Stampa…

Nisoldipine ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Isobutyl Methyl 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylate, 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic Acid Isobutyl Methyl Ester, 3-Isobutyl 5-methyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate

N0900-1G N0900-5G
TCIAN0900-1GEA 65 EUR
TCIAN0900-1G TCIAN0900-5G
Nisoldipine ≥98.0% (by HPLC, total nitrogen)
Nisoldipine
Formula: C₂₀H₂₄N₂O₆
Peso molecolare: 388,42 g/mol
MDL Number: MFCD00478055
CAS Number: 63675-72-9
EINECS: 264-407-7
Merck Index: 14,06565
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0d3574e4bb307e9dcd9e33b93fa7016b08c85cd&vid=d0d3574e4bb307e9dcd9e33b93fa7016b08c85cd&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 148.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR