Stampa…

Nilvadipine ≥95.0% (by total nitrogen basis)

Fornitore: TCI
Attenzione

Synonyms: 2-Cyano-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 5-Isopropyl 3-Methyl Ester, 5-Isopropyl 3-Methyl 2-Cyano-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate

N0899-1G N0899-5G
TCIAN0899-1GEA 90 EUR
TCIAN0899-1G TCIAN0899-5G
Nilvadipine ≥95.0% (by total nitrogen basis)
Nilvadipine
Formula: C₁₉H₁₉N₃O₆
Peso molecolare: 385.38 g/mol
MDL Number: MFCD00867065
CAS Number: 75530-68-6
Merck Index: 14,06545

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 149.0 to 153.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR