5-Norbornene-2-carbonitrile (mixture of isomers) ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Bicyclo[2.2.1]hept-5-ene-2-carbonitrile, 5-Norbornene-2-carbonitrile, 5-Cyanobicyclo[2.2.1]hept-2-ene, 5-Cyano-2-norbornene
N0897-25G
N0897-5G
TCIAN0897-25GEA
170
EUR
TCIAN0897-25G
TCIAN0897-5G
5-Norbornene-2-carbonitrile (mixture of isomers) ≥98.0% (by GC)
5-Norbornene-2-carbonitrile (mixture of isomers)
Formula:
C₈H₉N Peso molecolare: 119,17 g/mol Punto di fusione: 10°C Flash Pt: 66°C |
MDL Number:
MFCD00167563 CAS Number: 95-11-4 EINECS: 202-391-5 |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green clear liquid |
Purity (GC) | min. 98.0 % (total of isomer) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...