Stampa…

4-Nitro-L-phenylalanine methyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: H-Phe(4-NO2)-OMe.HCl, H-Phe(4-NO₂)-OMe·HCl

N0878-1G N0878-5G
TCIAN0878-1GEA 18 EUR
TCIAN0878-1G TCIAN0878-5G
4-Nitro-L-phenylalanine methyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Nitro-L-phenylalanine methyl ester hydrochloride
Formula: C₁₀H₁₂N₂O₄
Peso molecolare: 260,67 g/mol
Punto di fusione: 224°C
MDL Number: MFCD00135754
CAS Number: 17193-40-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +7.0 to +10.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR