Stampa…

Neu5Ac-α(2-6)Gal-β-MP glycoside ≥95.0% (by HPLC)

Fornitore: TCI

Synonyms: Neu5Ac α(2-6)Gal-β-MP, 4-Methoxyphenyl O-(5-Acetamido-3,5-dideoxy-D-glycero-α-D-galacto-2-nonulopyranosonic Acid)-(2-6)-β-D-galactopyranoside, 4-Methoxyphenyl 6-O-(N-Acetyl-α-neuraminosyl)-β-D-galactopyranoside

N0792-10MG N0792-50MG
TCIAN0792-10MGEA 132 EUR
TCIAN0792-10MG TCIAN0792-50MG
Neu5Ac-α(2-6)Gal-β-MP glycoside ≥95.0% (by HPLC)
Neu5Ac-alpha(2-6)Gal-beta-MP glycoside
Formula: C₂₄H₃₅NO₁₅
Peso molecolare: 577.54 g/mol
Temperatura di stoccaggio: Congelamento
CAS Number: 1984814-41-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR