Neopentyl glycol dimethacrylate ≥90.0% (by GC) stabilizzato
Fornitore: TCI
|
Attenzione
|
Synonyms:
2,2-Dimethylpropane-1,3-diyl bis(2-methylacrylate), 2,2-Dimethyl-1,3-propanediol Dimethacrylate, 1,3-Bis(methacryloyloxy)-2,2-dimethylpropane
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
Formula:
C₁₃H₂₀O₄ Peso molecolare: 240.30 g/mol |
MDL Number:
MFCD00048120 CAS Number: 1985-51-9 EINECS: 217-856-8 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...