3-(1-Naphthyl)-D-alanine hydrochloride ≥98.0% (by titrimetric analysis)
Fornitore: TCI
Formula:
C₁₃H₁₃NO₂ |
MDL Number:
MFCD03095597 CAS Number: 122745-09-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Specific rotation [a]20/D | +6.0 to +10.0 deg (C=1, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...