Stampa…

cis-5-Norbornene exo-2,3-dicarboxylic anhydride ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (3aR,4R,7S,7aS)-rel-3a,4,7,7a-Tetrahydro-4,7-methanoisobenzofuran-1,3-dione, Bicyclo[2.2.1]hept-5-ene-exo-2,3-dicarboxylic Anhydride

N0767-1G N0767-5G
TCIAN0767-1GEA 31 EUR
TCIAN0767-1G TCIAN0767-5G
cis-5-Norbornene exo-2,3-dicarboxylic anhydride ≥97.0% (by GC, titration analysis)
cis-5-Norbornene exo-2,3-dicarboxylic anhydride
Formula: C₉H₈O₃
Peso molecolare: 164,16 g/mol
MDL Number: MFCD00630580
CAS Number: 2746-19-2
EINECS: 220-384-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Methanolysis Method) min. 97.0 %
Melting point 142.0 to 146.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR