Stampa…

Naphthalene-1,4,5,8-tetracarboxylic dianhydride ≥98.0% (by HPLC), purified by sublimation

Fornitore: TCI EUROPE
Attenzione

Synonyms: NTCDA, Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride, 1,4,5,8-Naphthalenetetracarboxylic dianhydride

N0755-1G N0755-5G
TCIAN0755-1GEA 65 EUR
TCIAN0755-1G TCIAN0755-5G
Naphthalene-1,4,5,8-tetracarboxylic dianhydride ≥98.0% (by HPLC), purified by sublimation
Naphthalene-1,4,5,8-tetracarboxylic dianhydride
Formula: C₁₄H₄O₆
Peso molecolare: 268,18 g/mol
MDL Number: MFCD00006915
CAS Number: 81-30-1
EINECS: 201-342-5

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR