Stampa…

4-Nitro-L-phenylalanine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI

N0682-1G N0682-5G
TCIAN0682-1GEA 20 EUR
TCIAN0682-1G TCIAN0682-5G
4-Nitro-L-phenylalanine ≥98.0% (by HPLC, titration analysis)
4-Nitro-L-phenylalanine
Formula: C₉H₁₀N₂O₄
MDL Number: MFCD00150543
CAS Number: 949-99-5
EINECS: 213-446-8

Visualizza i codici

Risultati del test di specificazione

Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +5.0 to +7.0 deg (C=1, 1mol/L HCl)
Water max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR