Stampa…

Monoethyl-5-nitroisophthalate ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 5-Nitroisophthalic Acid Monoethyl Ester

N0676-25G N0676-500G
TCIAN0676-25GEA 32 EUR
TCIAN0676-25G TCIAN0676-500G
Monoethyl-5-nitroisophthalate ≥98.0% (by GC, titration analysis)
Monoethyl-5-nitroisophthalate
Formula: C₁₀H₉NO₆
Peso molecolare: 239,18 g/mol
MDL Number: MFCD00130051
CAS Number: 22871-55-2
EINECS: 245-273-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR