5-Norbornene-2-carboxylic acid (endo + exo mixture) ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Bicyclo[2.2.1]hept-5-ene-2-carboxylic Acid
N0667-25G
N0667-5G
TCIAN0667-25GEA
56
EUR
TCIAN0667-25G
TCIAN0667-5G
5-Norbornene-2-carboxylic acid (endo + exo mixture) ≥98.0% (by GC, titration analysis)
5-Norbornene-2-carboxylic acid
Formula:
C₈H₁₀O₂ Peso molecolare: 138,17 g/mol |
MDL Number:
MFCD00085356 CAS Number: 120-74-1 EINECS: 204-422-8 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % (total of isomer) |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...