Dimethyl-2,3-naphthalenedicarboxylate ≥97.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
2,3-Naphthalenedicarboxylic Acid Dimethyl Ester
N0590-1G
N0590-25G
N0590-5G
TCIAN0590-1GEA
48
EUR
TCIAN0590-1G
TCIAN0590-25G
TCIAN0590-5G
Dimethyl-2,3-naphthalenedicarboxylate ≥97.0% (by GC)
Dimethyl-2,3-naphthalenedicarboxylate
Formula:
C₁₄H₁₂O₄ Peso molecolare: 244,25 g/mol Punto di ebollizione: 145°C |
MDL Number:
MFCD00012278 CAS Number: 13728-34-2 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 97.0 % |
Melting point | 48.0 to 52.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...