Nifedipine ≥98.0% (by HPLC and UV method)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Adalat, 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic Acid Dimethyl Ester
N0528-10G
N0528-25G
TCIAN0528-10GEA
59
EUR
TCIAN0528-10G
TCIAN0528-25G
Nifedipine ≥98.0% (by HPLC and UV method)
Nifedipine
Formula:
C₁₇H₁₈N₂O₆ Peso molecolare: 346,34 g/mol |
MDL Number:
MFCD00057326 CAS Number: 21829-25-4 EINECS: 244-598-3 Merck Index: 14,06528 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (UV-vis method) | min. 98.0 % |
Melting point | 172.0 to 175.0 deg-C |
Solubility in Acetone | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...