Stampa…

syn-4-Nitrobenzaldehyde oxime ≥85.0% (by GC) deprotecting agent

Fornitore: TCI EUROPE
Attenzione

Synonyms: syn-4-Nitrobenzaldoxime

N0487-25G N0487-5G
TCIAN0487-25GEA 98 EUR
TCIAN0487-25G TCIAN0487-5G
syn-4-Nitrobenzaldehyde oxime ≥85.0% (by GC) deprotecting agent
syn-4-Nitrobenzaldehyde oxime
Formula: C₇H₆N₂O₃
Peso molecolare: 166,14 g/mol
MDL Number: MFCD00151329
CAS Number: 3717-19-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c67d8f6e881eb7efe64cb2dff48d7db503eac5a&vid=0c67d8f6e881eb7efe64cb2dff48d7db503eac5a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 85.0 %
Melting point 124.0 to 128.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR