Nitrilotris(methylenephosphonic acid) ∼50% ∼2.2 mol/L in water
Fornitore: TCI
|
Attenzione
|
Formula:
C₃H₁₂NO₉P₃ |
MDL Number:
MFCD00002138 CAS Number: 6419-19-8 EINECS: 229-146-5 |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Concentration (Neutralization titration) | 47.0 to 53.0 w/w% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...