Neopentyl glycol mono(hydroxypivalate) ≥97.0% (by GC)
Fornitore: TCI
Synonyms:
2,2-Dimethyl-1,3-propanediol Mono(hydroxypivalate), 3-Hydroxy-2,2-dimethylpropyl 3-Hydroxy-2,2-dimethylpropionate, Hydroxypivalic Acid Neopentyl Glycol Ester
Formula:
C₁₀H₂₀O₄ Peso molecolare: 204.27 g/mol |
MDL Number:
MFCD00059597 CAS Number: 1115-20-4 EINECS: 214-222-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 51.0 to 55.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...