Stampa…
Neopentyl glycol diglycidyl Ether 40,00% (by GC)
Fornitore: TCI
Attenzione
Synonyms: Diglycidyloxyneopentane, Neopentyl glycol diglycidyl ether
N0448-25ML N0448-500ML
TCIAN0448-25MLEA 16 EUR
TCIAN0448-25ML TCIAN0448-500ML
Neopentyl glycol diglycidyl Ether 40,00% (by GC)
Neopentyl glycol diglycidyl Ether
Formula: C₁₁H₂₀O₄
Peso molecolare: 216.28 g/mol
Flash Pt: 141°C
MDL Number: MFCD00040687
CAS Number: 17557-23-2
EINECS: 241-536-7
Visualizza i codici
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 40.0 %