Stampa…

Sodium naphthenate

Fornitore: TCI

Synonyms: Naphthenic Acid Sodium Salt

N0397-25G N0397-500G
TCIAN0397-25GEA 60 EUR
TCIAN0397-25G TCIAN0397-500G
Sodium naphthenate
Sodium naphthenate
MDL Number: MFCD00147698
CAS Number: 61790-13-4
EINECS: 263-108-9
UN: 3077
ADR: 9 III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Sodium 8.0 to 12.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR