Stampa…

Neostigmine bromide ≥99.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 3-((Dimethylcarbamoyl)oxy)-N,N,N-trimethylbenzenaminium bromide, (3-Dimethylcarbamoyloxyphenyl)trimethylammonium Bromide, (3-Hydroxyphenyl)trimethylammonium Bromide Dimethyl Carbamate

N0358-25G N0358-5G
TCIAN0358-25GEA 162 EUR
TCIAN0358-25G TCIAN0358-5G
Neostigmine bromide ≥99.0% (by titrimetric analysis)
Neostigmine bromide
Formula: C₁₂H₁₉BrN₂O₂
Peso molecolare: 303,20 g/mol
MDL Number: MFCD00011795
CAS Number: 114-80-7
EINECS: 204-054-8
UN: 2811
ADR: 6.1,III
Merck Index: 14,06464

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 % (after drying)
Solubility in Water almost transparency
Drying loss max. 1.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR