Stampa…

Nonenylsuccinic anhydride (mixture of branched chain isomers) ≥90.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: NSA, Isononenylsuccinic anhydride

N0350-500G N0350-25G
TCIAN0350-500GEA 180 EUR
TCIAN0350-500G TCIAN0350-25G
Nonenylsuccinic anhydride (mixture of branched chain isomers) ≥90.0% (by titrimetric analysis)
Nonenylsuccinic anhydride (mixture of branched chain isomers)
Formula: C₁₃H₂₀O₃
Peso molecolare: 224,30 g/mol
Flash Pt: 147°C
MDL Number: MFCD00014552
CAS Number: 28928-97-4
EINECS: 249-317-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green clear liquid
Purity (Morpholine Method) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR