Naphthenic acid
Fornitore: TCI
|
Attenzione
|
Flash Pt:
165°C |
MDL Number:
MFCD00147696 CAS Number: 1338-24-5 EINECS: 215-662-8 UN: 3082 ADR: 9,III |
Risultati del test di specificazione
Appearance | Light yellow to Brown clear liquid |
Neutralization Value | 190.0 to 230.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...